The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_111689.5 | NP_187467.1 | 6 - 7 | 1370 | 120 |
GenBank mRNA | AF543710.1 | - | 6 - 7 | 892 | 120 |