Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACBP3
Entrez Gene ID
828524
Gene Name
acyl-CoA-binding domain 3
Gene Aliases
AT4G24230, T22A6.60, T22A6_60, acyl-CoA-binding domain 3
Chromosome Location
Chr.4: 12566631 - 12568866 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084970.1 NP_001078439.1 1 - 2 880 78
NM_001084971.1 NP_001078440.1 1 - 2 877 78
NM_001084972.2 NP_001078441.1 1 - 2 878 78
NM_001125569.1 NP_001119041.1 1 - 2 864 78
NM_118556.4 NP_194154.1 1 - 2 956 78
NM_179101.3 NP_849432.1 1 - 2 976 78
GenBank mRNA AK227063.1 - 1 - 2 932 78
AK317000.1 - 1 - 2 987 78

View Full Product Details