Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PWD
Entrez Gene ID
828547
Gene Name
phosphoglucan, water dikinase
Gene Aliases
AT4G24450, ATGWD2, GWD3, T22A6.280, T22A6_280, phosphoglucan, water dikinase
Chromosome Location
Chr.4: 12635061 - 12642808 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341673.1 NP_001328878.1 16 - 17 2183 118
NM_001341674.1 NP_001328879.1 16 - 17 2187 118
NM_118578.5 NP_194176.3 15 - 16 1985 118

View Full Product Details