Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATGP4
Entrez Gene ID
828602
Gene Name
Ubiquitin family protein
Gene Aliases
AT4G24990, F13M23.130, F13M23_130
Chromosome Location
Chr.4: 12849651 - 12851810 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341720.1 NP_001329044.1 2 - 3 820 74
NM_118631.3 NP_194229.1 3 - 4 765 74
GenBank mRNA AK227315.1 - 3 - 4 588 74
BT003091.1 - 2 - 3 300 74

View Full Product Details