Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACP4
Entrez Gene ID
828608
Gene Name
acyl carrier protein 4
Gene Aliases
AT4G25050, F24A6.4, acyl carrier protein 4
Chromosome Location
Chr.4: 12870006 - 12871564 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203902.1 NP_001190831.1 2 - 3 323 80
NM_118637.4 NP_194235.1 2 - 3 321 80
GenBank mRNA AY039527.1 - 2 - 3 214 80
AY050471.1 - 2 - 3 151 80
AY084704.1 - 2 - 3 262 80

View Full Product Details