Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRS2
Entrez Gene ID
828615
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
ARABIDOPSIS THALIANA SUPPRESSOR OF RAD SIX-SCREEN MUTANT 2, AT4G25120, ATSRS2, F24A6.8, SUPPRESSOR OF RAD SIX-SCREEN MUTANT 2
Chromosome Location
Chr.4: 12890545 - 12898501 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341722.1 NP_001320059.1 14 - 15 2321 82
GenBank mRNA GQ148553.1 - 14 - 15 2228 82

View Full Product Details