Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACD1-LIKE
Entrez Gene ID
828670
Gene Name
ACD1-like protein
Gene Aliases
52 KDA, 55 KDA - IV, ACD1-like, AT4G25650, L73G19.30, L73G19_30, PROTOCHLOROPHYLLIDE-DEPENDENT TRANSLOCON COMPONENT, PTC52, TIC55-IV, TRANSLOCON AT THE INNER ENVELOPE MEMBRANE OF CHLOROPLASTS
Chromosome Location
Chr.4: 13080827 - 13083260 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118697.4 NP_567725.1 2 - 3 808 102
NM_179113.2 NP_849444.1 2 - 3 745 102
GenBank mRNA AF424575.1 - 2 - 3 714 102
AY056168.1 - 2 - 3 737 102
AY062731.1 - 2 - 3 745 102
AY344062.1 - 2 - 3 701 102
BT002586.1 - 2 - 3 701 102

View Full Product Details