Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CPSUFE
- Entrez Gene ID
-
828756
- Gene Name
- chloroplast sulfur E
- Gene Aliases
-
ARABIDOPSIS THALIANA SULFUR E, AT4G26500, ATSUFE, EMB1374, M3E9.70, M3E9_70, MBRYO DEFECTIVE 1374, SUFE1, SULFUR E 1, chloroplast sulfur E
- Chromosome Location
-
Chr.4: 13381714 - 13383641 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118783.3 |
NP_194380.1 |
1 - 1 |
1359 |
124 |
- Gene Symbol
- UKL4
- Entrez Gene ID
-
828757
- Gene Name
- uridine kinase-like 4
- Gene Aliases
-
AT4G26510, M3E9.60, M3E9_60, uridine kinase-like 4
- Chromosome Location
-
Chr.4: 13381687 - 13388362 on Build TAIR10.1
View Full Product Details