Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY20
Entrez Gene ID
828771
Gene Name
WRKY family transcription factor family protein
Gene Aliases
AT4G26640, AtWRKY20, T15N24.90, T15N24_90
Chromosome Location
Chr.4: 13437005 - 13441884 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341821.1 NP_001329242.1 3 - 4 1648 78
NM_001341822.1 NP_001329238.1 2 - 3 1537 78
NM_001341823.1 NP_001329241.1 4 - 5 1493 78
NM_001341824.1 NP_001329240.1 3 - 4 1248 78
NM_001341825.1 NP_001329239.1 5 - 6 1692 78
NM_118798.2 NP_567752.1 4 - 5 1552 78
NM_179119.5 NP_849450.1 4 - 5 1356 78
GenBank mRNA AF425837.1 - 4 - 5 1053 78
AJ630479.1 - 4 - 5 1053 78
AY150436.1 - 3 - 4 837 78
AY568651.1 - 4 - 5 1053 78

View Full Product Details