Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FIM1
Entrez Gene ID
828777
Gene Name
fimbrin 1
Gene Aliases
ARABIDOPSIS THALIANA FIMBRIN 1, AT4G26700, ATFIM1, F10M23.40, F10M23_40, FIMBRIN-LIKE PROTEIN, fimbrin 1
Chromosome Location
Chr.4: 13463097 - 13467604 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036649.1 NP_001031726.1 11 - 12 1781 125
NM_001036650.2 NP_001031727.1 10 - 11 1868 125
NM_001341826.1 NP_001328753.1 11 - 12 1908 125
NM_001341827.1 NP_001328754.1 11 - 12 1892 125
NM_001341828.1 NP_001328752.1 11 - 12 1880 125
NM_001341829.1 NP_001328755.1 9 - 10 1802 125
NM_118804.5 NP_194400.1 10 - 11 1732 125
GenBank mRNA AK318682.1 - 11 - 12 1784 125

View Full Product Details