Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCG38
Entrez Gene ID
822863
Gene Name
pleiotropic drug resistance 10
Gene Aliases
AT3G30842, ATP-binding cassette G38, ATPDR10, MJI6.16, PDR10, pleiotropic drug resistance 10
Chromosome Location
Chr.3: 12593810 - 12600432 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339062.1 NP_001319673.1 5 - 6 763 63

View Full Product Details