Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP71A25
- Entrez Gene ID
-
823986
- Gene Name
- cytochrome P450, family 71, subfamily A, polypeptide 25
- Gene Aliases
-
''cytochrome P450, AT3G48280, T29H11.200, cytochrome P450, family 71, polypeptide 25, polypeptide 25'', subfamily A
- Chromosome Location
-
Chr.3: 17879542 - 17881388 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_148854.3 |
NP_680107.1 |
1 - 2 |
909 |
65 |
View Full Product Details