Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nek7
Entrez Gene ID
820397
Gene Name
NIMA-related kinase 7
Gene Aliases
AT3G12200, AtNek7, NIMA-related kinase 7
Chromosome Location
Chr.3: 3886957 - 3891000 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202938.1 NP_001189867.1 11 - 12 1146 110
NM_001337978.1 NP_001327416.1 9 - 10 1193 110
NM_001337979.1 NP_001327417.1 10 - 11 1407 110
NM_112055.5 NP_187827.1 10 - 11 1047 110
GenBank mRNA BT015419.1 - 10 - 11 666 110

View Full Product Details