Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nek7
Entrez Gene ID
820397
Gene Name
NIMA-related kinase 7
Gene Aliases
AT3G12200, AtNek7, NIMA-related kinase 7
Chromosome Location
Chr.3: 3886957 - 3891000 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112055.5 NP_187827.1 3 - 4 648 124
GenBank mRNA BT015419.1 - 3 - 4 267 124

View Full Product Details