Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nek7
Entrez Gene ID
820397
Gene Name
NIMA-related kinase 7
Gene Aliases
AT3G12200, AtNek7, NIMA-related kinase 7
Chromosome Location
Chr.3: 3886957 - 3891000 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202938.1 NP_001189867.1 9 - 10 1012 82
NM_001337978.1 NP_001327416.1 7 - 8 1059 82
NM_001337979.1 NP_001327417.1 8 - 9 1273 82
NM_112055.5 NP_187827.1 8 - 9 913 82
GenBank mRNA BT015419.1 - 8 - 9 532 82

View Full Product Details