Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rps15ae
Entrez Gene ID
829064
Gene Name
ribosomal protein S15A E
Gene Aliases
AT4G29430, F17A13.250, F17A13_250, ribosomal protein S15A E
Chromosome Location
Chr.4: 14472303 - 14473621 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119088.4 NP_194672.1 1 - 2 262 111
GenBank mRNA AK228983.1 - 1 - 2 158 111
AY085780.1 - 1 - 2 163 111
BT026503.1 - 1 - 2 125 111

View Full Product Details