Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRHA
Entrez Gene ID
829117
Gene Name
pathogenesis related homeodomain protein A
Gene Aliases
AT4G29940, F27B13.180, F27B13_180, HOMEODOMAIN PROTEIN, pathogenesis related homeodomain protein A, pathogenesis related homeodomain protein A
Chromosome Location
Chr.4: 14648138 - 14653489 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203938.2 NP_001190867.1 7 - 8 1719 67
NM_119140.2 NP_194723.1 7 - 8 1695 67
GenBank mRNA L21991.1 - 7 - 8 1610 67

View Full Product Details