Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMA2
Entrez Gene ID
829134
Gene Name
heavy metal atpase 2
Gene Aliases
ARABIDOPSIS HEAVY METAL ATPASE 2, AT4G30110, ATHMA2, F6G3.140, F6G3_140, heavy metal atpase 2
Chromosome Location
Chr.4: 14720062 - 14725311 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341993.1 NP_001320088.1 5 - 6 949 68
NM_119157.4 NP_194740.1 4 - 5 804 68

View Full Product Details