Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AT4G30380
Entrez Gene ID
829161
Gene Name
Barwin-related endoglucanase
Gene Aliases
AT4G30380, EXLB2, EXPANSIN-LIKE B2 PRECURSOR, F17I23.280, F17I23_280
Chromosome Location
Chr.4: 14860480 - 14861239 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342007.1 NP_001320090.1 1 - 2 96 75

View Full Product Details