Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl29
Entrez Gene ID
829204
Gene Name
serine carboxypeptidase-like 29
Gene Aliases
AT4G30810, F6I18.280, F6I18_280, serine carboxypeptidase-like 29
Chromosome Location
Chr.4: 15003146 - 15006216 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119227.4 NP_567854.1 9 - 10 1579 75
GenBank mRNA AY050958.1 - 9 - 10 1274 75
AY133774.1 - 9 - 10 1251 75

View Full Product Details