Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP24
Entrez Gene ID
829213
Gene Name
ubiquitin-specific protease 24
Gene Aliases
AT4G30890, F6I18.200, F6I18_200, ubiquitin-specific protease 24
Chromosome Location
Chr.4: 15036020 - 15039417 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203946.1 NP_001190875.1 6 - 7 1671 150
NM_119236.4 NP_567860.1 6 - 7 1641 150
NM_202915.2 NP_974644.1 5 - 6 1884 150
GenBank mRNA AF302672.1 - 5 - 6 1307 150

View Full Product Details