Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY32
Entrez Gene ID
829218
Gene Name
WRKY DNA-binding protein 32
Gene Aliases
AT4G30935, ATWRKY32, F6I18.160, F6I18_160, WRKY DNA-binding protein 32
Chromosome Location
Chr.4: 15051764 - 15054107 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119241.6 NP_567862.3 4 - 5 1248 66
GenBank mRNA BT004086.1 - 4 - 5 1106 66
BT005110.1 - 4 - 5 1049 66

View Full Product Details