Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRE11
Entrez Gene ID
835514
Gene Name
DNA repair and meiosis protein (Mre11)
Gene Aliases
ARABIDOPSIS MEIOTIC RECOMBINATION 11, AT5G54260, ATMRE11, MDK4.8, MDK4_8, MEIOTIC RECOMBINATION 11
Chromosome Location
Chr.5: 22032038 - 22038052 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345081.1 NP_001330550.1 2 - 3 318 68
NM_124806.2 NP_200237.1 4 - 5 504 68
GenBank mRNA AJ243822.1 - 4 - 5 473 68

View Full Product Details