Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGD5
Entrez Gene ID
835519
Gene Name
ARF-GAP domain 5
Gene Aliases
ARF-GAP domain 5, AT5G54310, AtAGD5, MDK4.13, MDK4_13, NEV, NEVERSHED
Chromosome Location
Chr.5: 22056986 - 22061270 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124811.5 NP_568807.1 4 - 5 458 91
GenBank mRNA AK226402.1 - 4 - 5 266 91
AY099716.1 - 4 - 5 404 91
BT000287.1 - 4 - 5 254 91
FJ794601.1 - 4 - 5 356 91

View Full Product Details