Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATK3
Entrez Gene ID
835556
Gene Name
kinesin 3
Gene Aliases
AT5G54670, KATC, KINESIN-LIKE PROTEIN IN ARABIDOPSIS THALIANA C, MRB17.18, kinesin 3
Chromosome Location
Chr.5: 22209403 - 22214252 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345094.1 NP_001330580.1 12 - 13 1813 115
NM_001345095.1 NP_001330581.1 10 - 11 1437 115
NM_124848.3 NP_568811.1 12 - 13 1932 115
GenBank mRNA AK228138.1 - 12 - 13 1863 115
BT003156.1 - 11 - 12 1584 115
D21138.1 - 12 - 13 1834 115

View Full Product Details