Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPX2
Entrez Gene ID
835619
Gene Name
protein phosphatase X 2
Gene Aliases
AT5G55260, MCO15.21, MCO15_21, PPX-2, PROTEIN PHOSPHATASE X -2, protein phosphatase X 2
Chromosome Location
Chr.5: 22416578 - 22418990 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345121.1 NP_001330655.1 5 - 6 1135 125
NM_124908.4 NP_200337.1 6 - 7 1010 125
GenBank mRNA BT024716.1 - 6 - 7 782 125

View Full Product Details