Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOP1ALPHA
Entrez Gene ID
835623
Gene Name
DNA topoisomerase I alpha
Gene Aliases
AT5G55300, DNA topoisomerase 1 beta, DNA topoisomerase I alpha, MGO1, MGOUN 1, MTE17.1, MTE17_1, TOP1, TOP1BETA, TOPOISOMERASE 1
Chromosome Location
Chr.5: 22424502 - 22429514 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203614.1 NP_001190543.1 10 - 11 2256 134
NM_001203615.1 NP_001190544.1 4 - 5 1193 134
NM_124912.5 NP_200341.1 10 - 11 2268 134
GenBank mRNA X57544.1 - - 2193 134

View Full Product Details