Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TEX1
Entrez Gene ID
835712
Gene Name
Transducin/WD40 repeat-like superfamily protein
Gene Aliases
AT5G56130, AtTEX1, MDA7.19, MDA7_19, THO3
Chromosome Location
Chr.5: 22722482 - 22725099 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_124995.5 NP_200424.1 4 - 5 417 84
GenBank mRNA AY093120.1 - 4 - 5 403 84
BT000174.1 - 4 - 5 383 84

View Full Product Details