Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WRKY20
Entrez Gene ID
828771
Gene Name
WRKY family transcription factor family protein
Gene Aliases
AT4G26640, AtWRKY20, T15N24.90, T15N24_90
Chromosome Location
Chr.4: 13437005 - 13441884 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341821.1 NP_001329242.1 1 - 2 536 94
NM_001341823.1 NP_001329241.1 1 - 2 536 94
NM_001341825.1 NP_001329239.1 2 - 3 872 94
NM_179119.5 NP_849450.1 1 - 2 536 94
GenBank mRNA AF425837.1 - 1 - 2 233 94
AJ630479.1 - 1 - 2 233 94
AY568651.1 - 1 - 2 233 94

View Full Product Details