Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGT73B2
Entrez Gene ID
829560
Gene Name
UDP-glucosyltransferase 73B2
Gene Aliases
AT4G34135, UDP-glucosyltransferase 73B2
Chromosome Location
Chr.4: 16345150 - 16347434 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179161.2 NP_849492.1 1 - 1 1418 103

View Full Product Details