Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMT2
Entrez Gene ID
829684
Gene Name
tonoplast monosaccharide transporter2
Gene Aliases
AT4G35300, F23E12.140, F23E12_140, tonoplast monosaccharide transporter2
Chromosome Location
Chr.4: 16795775 - 16799823 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160815.2 NP_001154287.1 2 - 3 1208 75
NM_001203993.1 NP_001190922.1 4 - 5 719 75
NM_001203994.1 NP_001190923.1 3 - 4 838 75
NM_001342339.1 NP_001328125.1 3 - 4 950 75
NM_001342340.1 NP_001328126.1 3 - 4 886 75
NM_001342341.1 NP_001328129.1 3 - 4 1093 75
NM_001342342.1 NP_001328130.1 2 - 3 1384 75
NM_001342343.1 NP_001328127.1 3 - 4 1118 75
NM_001342344.1 NP_001328128.1 3 - 4 1086 75
NM_119696.5 NP_195256.3 3 - 4 951 75
NM_179234.2 NP_849565.1 3 - 4 920 75
GenBank mRNA AJ532570.1 - 2 - 3 644 75
AY094465.1 - 3 - 4 852 75

View Full Product Details