Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IMPA-4
Entrez Gene ID
837448
Gene Name
importin alpha
Gene Aliases
AT1G09270, T12M4.2, T12M4_2, importin alpha isoform 4
Chromosome Location
Chr.1: 2994331 - 2998222 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035934.2 NP_001031011.1 5 - 6 833 110
NM_100797.6 NP_172398.1 5 - 6 920 110
NM_179292.3 NP_849623.1 5 - 6 920 110
GenBank mRNA AK316689.1 - 5 - 6 817 110
AY090944.1 - 5 - 6 843 110
AY117313.1 - 5 - 6 745 110

View Full Product Details