Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IMPA-4
Entrez Gene ID
837448
Gene Name
importin alpha
Gene Aliases
AT1G09270, T12M4.2, T12M4_2, importin alpha isoform 4
Chromosome Location
Chr.1: 2994331 - 2998222 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035934.2 NP_001031011.1 7 - 8 1066 69
NM_100797.6 NP_172398.1 7 - 8 1153 69
NM_179292.3 NP_849623.1 7 - 8 1153 69
GenBank mRNA AK316689.1 - 7 - 8 1050 69
AY090944.1 - 7 - 8 1076 69
AY117313.1 - 7 - 8 978 69

View Full Product Details