Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACLA-1
Entrez Gene ID
837610
Gene Name
ATP-citrate lyase A-1
Gene Aliases
AT1G10670, ATP-citrate lyase A-1, F20B24.11, F20B24_11
Chromosome Location
Chr.1: 3535288 - 3538439 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198024.1 NP_001184953.1 8 - 9 956 105
NM_001198025.1 NP_001184954.1 7 - 8 963 105
NM_100943.4 NP_172537.1 7 - 8 991 105
NM_179303.4 NP_849634.1 6 - 7 919 105
GenBank mRNA AK230271.1 - - 1756 105
AY056213.1 - 7 - 8 746 105
AY056593.1 - 7 - 8 737 105
AY094479.1 - 7 - 8 749 105
AY113979.1 - 6 - 7 687 105

View Full Product Details