Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBP45B
Entrez Gene ID
837708
Gene Name
RNA-binding (RRM/RBD/RNP motifs) family protein
Gene Aliases
AT1G11650, ATRBP45B, F25C20.21, F25C20_21
Chromosome Location
Chr.1: 3914774 - 3918243 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101037.4 NP_172630.1 1 - 2 385 68
NM_179310.2 NP_849641.1 1 - 2 385 68
GenBank mRNA AY093201.1 - 1 - 2 356 68
BT008494.1 - 1 - 2 264 68

View Full Product Details