Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ML5
Entrez Gene ID
839816
Gene Name
MEI2-like protein 5
Gene Aliases
AML5, AT1G29400, F15D2.30, F15D2_30, MEI2-like protein 5
Chromosome Location
Chr.1: 10289998 - 10294630 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332851.1 NP_001320501.1 3 - 4 1081 109
NM_102680.2 NP_174233.2 3 - 4 689 109
NM_179396.3 NP_849727.1 3 - 4 838 109
GenBank mRNA AY062613.1 - 3 - 4 725 109
AY070368.1 - 3 - 4 728 109
AY091327.1 - 2 - 3 565 109

View Full Product Details