Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALIS1
Entrez Gene ID
820456
Gene Name
ALA-interacting subunit 1
Gene Aliases
ALA-interacting subunit 1, AT3G12740
Chromosome Location
Chr.3: 4049309 - 4051775 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112110.4 NP_566435.1 6 - 7 1141 68
GenBank mRNA AF410309.1 - 6 - 7 770 68
AY045904.1 - 6 - 7 846 68
AY087607.1 - 6 - 7 935 68
AY091210.1 - 6 - 7 711 68

View Full Product Details