Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRF5
Entrez Gene ID
820609
Gene Name
growth-regulating factor 5
Gene Aliases
AT3G13960, AtGRF5, growth-regulating factor 5
Chromosome Location
Chr.3: 4608076 - 4610497 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112250.3 NP_188012.2 3 - 4 1633 82
GenBank mRNA AY102638.1 - 3 - 4 1326 82

View Full Product Details