Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATBRXL2
Entrez Gene ID
820614
Gene Name
DZC (Disease resistance/zinc finger/chromosome condensation-like region) domain containing protein
Gene Aliases
AT3G14000, BRX-LIKE2
Chromosome Location
Chr.3: 4630865 - 4634081 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112254.3 NP_188016.2 3 - 4 719 74
NM_202580.2 NP_974309.1 4 - 5 1292 74
GenBank mRNA AK117522.1 - 3 - 4 716 74
BT029002.1 - 3 - 4 658 74

View Full Product Details