Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ESM1
Entrez Gene ID
820639
Gene Name
GDSL-like lipase/acylhydrolase superfamily protein
Gene Aliases
AT3G14210, epithiospecifier modifier 1
Chromosome Location
Chr.3: 4729399 - 4731847 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338114.1 NP_001326872.1 3 - 4 527 100
NM_112278.3 NP_188037.1 2 - 3 869 100
GenBank mRNA AK221834.1 - 2 - 3 445 100
AY054230.1 - 2 - 3 423 100
AY062690.1 - 2 - 3 425 100
BT002522.1 - 2 - 3 425 100
BT008484.1 - 2 - 3 382 100

View Full Product Details