Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPK19
Entrez Gene ID
820700
Gene Name
MAP kinase 19
Gene Aliases
ARABIDOPSIS THALIANA MAP KINASE 19, AT3G14720, ATMPK19, MAP kinase 19
Chromosome Location
Chr.3: 4945601 - 4949042 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338134.1 NP_001326968.1 6 - 7 1386 135
NM_001338135.1 NP_001326969.1 6 - 7 1386 135
NM_112333.3 NP_188090.2 6 - 7 1386 135

View Full Product Details