Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CESA1
Entrez Gene ID
829376
Gene Name
cellulose synthase 1
Gene Aliases
ANY1, AT4G32410, AtCESA1, CELLULOSE SYNTHASE 1, F8B4.110, F8B4_110, RADIALLY SWOLLEN 1, RSW1, anisotropy1, cellulose synthase 1
Chromosome Location
Chr.4: 15640490 - 15646844 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119393.3 NP_194967.1 7 - 8 1822 133
GenBank mRNA AK226243.1 - 7 - 8 1501 133
BT008654.1 - 7 - 8 1510 133
KJ138727.1 - 7 - 8 1366 133

View Full Product Details