Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KAT3
Entrez Gene ID
829400
Gene Name
potassium channel protein
Gene Aliases
A. thaliana low-K+-tolerant 1, ARABIDOPSIS THALIANA K+ RECTIFYING CHANNEL 1, AT4G32650, ATKC1, AtLKT1, F4D11.150, F4D11_150, K+ INWARD RECTIFYING CHANNEL PROTEIN, KC1, potassium channel in Arabidopsis thaliana 3
Chromosome Location
Chr.4: 15751271 - 15755047 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036696.1 NP_001031773.1 8 - 9 1424 134
NM_119417.4 NP_194991.1 8 - 9 1626 134
NM_202936.1 NP_974665.1 8 - 9 1433 134
GenBank mRNA AY052233.1 - 8 - 9 1424 134
BT003093.1 - 8 - 9 1376 134
U81239.1 - 8 - 9 1381 134
Z83202.1 - - 1422 134

View Full Product Details