Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PC-MYB1
Entrez Gene ID
829409
Gene Name
Homeodomain-like protein
Gene Aliases
A. thaliana MYB domain protein 3R1, ARABIDOOSIS THALIANA MYB DOMAIN PROTEIN 3R1, AT4G32730, ATMYB3R-1, ATMYB3R1, C-MYB-LIKE TRANSCRIPTION FACTOR 3R-1, F4D11.70, F4D11_70, MYB DOMAIN PROTEIN 3R1, MYB3R-1, MYB3R1
Chromosome Location
Chr.4: 15790133 - 15796114 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342169.1 NP_001328944.1 4 - 5 715 110
NM_001342170.1 NP_001328945.1 4 - 5 503 110
NM_001342171.1 NP_001328946.1 4 - 5 488 110
NM_119426.2 NP_194999.1 4 - 5 488 110
NM_202989.3 NP_974718.1 4 - 5 700 110
GenBank mRNA AF151646.1 - 3 - 4 180 110
AF176005.2 - 4 - 5 488 110

View Full Product Details