Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AUR1
Entrez Gene ID
829419
Gene Name
ataurora1
Gene Aliases
AT4G32830, AtAUR1, T16I18.40, T16I18_40, ataurora1
Chromosome Location
Chr.4: 15842443 - 15844865 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119436.3 NP_195009.1 1 - 2 152 92
GenBank mRNA AJ854183.1 - 1 - 2 38 92
AK221608.1 - 1 - 2 142 92

View Full Product Details