Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFK6
Entrez Gene ID
829420
Gene Name
phosphofructokinase 6
Gene Aliases
AT4G32840, T16I18.50, T16I18_50, phosphofructokinase 6
Chromosome Location
Chr.4: 15844576 - 15848552 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119437.3 NP_195010.1 1 - 2 352 71
GenBank mRNA AK227471.1 - 1 - 2 241 71
BT006001.1 - 1 - 2 105 71

View Full Product Details