Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HB-8
Entrez Gene ID
829424
Gene Name
homeobox-leucine zipper protein ATHB-8
Gene Aliases
AT4G32880, ATHB-8, ATHB8, T16I18.90, T16I18_90, homeobox gene 8
Chromosome Location
Chr.4: 15863339 - 15868650 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119441.5 NP_195014.1 6 - 7 1067 66
GenBank mRNA AJ441292.1 - 5 - 6 656 66
AY099631.1 - 6 - 7 1042 66
BT008798.1 - 5 - 6 656 66

View Full Product Details