Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APM1
Entrez Gene ID
829446
Gene Name
aminopeptidase M1
Gene Aliases
AMINOPEPTIDASE M1, AT4G33090, ATAPM1, F4I10.20, F4I10_20, aminopeptidase M1
Chromosome Location
Chr.4: 15965593 - 15970557 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119463.5 NP_195035.2 5 - 6 888 102
GenBank mRNA AK227105.1 - 5 - 6 785 102
AY064961.1 - 5 - 6 786 102
AY143574.1 - 5 - 6 761 102
BT005811.1 - 5 - 6 749 102

View Full Product Details