Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APM1
Entrez Gene ID
829446
Gene Name
aminopeptidase M1
Gene Aliases
AMINOPEPTIDASE M1, AT4G33090, ATAPM1, F4I10.20, F4I10_20, aminopeptidase M1
Chromosome Location
Chr.4: 15965593 - 15970557 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119463.5 NP_195035.2 8 - 9 1261 97
GenBank mRNA AK227105.1 - 8 - 9 1158 97
AY064961.1 - 8 - 9 1159 97
AY143574.1 - 8 - 9 1134 97
BT005811.1 - 8 - 9 1122 97

View Full Product Details