Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DML3
Entrez Gene ID
829552
Gene Name
demeter-like protein 3
Gene Aliases
AT4G34060, F28A23.180, F28A23_180, demeter-like protein 3
Chromosome Location
Chr.4: 16313366 - 16319426 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125640.1 NP_001119112.1 4 - 5 886 90
NM_001342250.1 NP_001329149.1 5 - 6 1113 90
NM_001342251.1 NP_001329150.1 2 - 3 1138 90
NM_001342252.1 NP_001329148.1 3 - 4 986 90
NM_119567.4 NP_195132.3 4 - 5 1040 90

View Full Product Details